Learn More
(R)-(-)-2-Amino-1-butanol, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 244031000
| Quantity | 100mL |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C4H11NO | |
| MFCD00064419 | |
| r-2-aminobutan-1-ol, r---2-amino-1-butanol, 2r-2-aminobutan-1-ol, r-2-amino-1-butanol, 1-butanol, 2-amino-, 2r, unii-4f725v55ln, 2-amino-1-butanol, r, 2-amino-1-butanol,-, l---2-amino-1-butanol, 1-butanol, 2-amino-, r | |
| 2-aminobutan-1-ol |
Specifications
| (R)-(-)-2-Amino-1-butanol | |
| 97.5 | |
| 0.9470g/mL | |
| 90°C | |
| 97.5% min. (GC) | |
| C4H11NO | |
| C2H5CH(NH2)CH2OH | |
| MFCD00064419 | |
| 0.947 | |
| -10 | |
| Solubility in water: miscible. Other solubilities: very soluble in most of the usual solvents,hardly soluble in aliphatic hydrocarbons | |
| CCC(N)CO | |
| 89.14 | |
| 89.14 |
| 5856-63-3 | |
| 100.0 | |
| 172°C to 174°C | |
| Authentic | |
| Glass bottle | |
| 1.4495 to 1.4535 | |
| 100mL | |
| 04,III,771 | |
| 10, 429 | |
| r-2-aminobutan-1-ol, r---2-amino-1-butanol, 2r-2-aminobutan-1-ol, r-2-amino-1-butanol, 1-butanol, 2-amino-, 2r, unii-4f725v55ln, 2-amino-1-butanol, r, 2-amino-1-butanol,-, l---2-amino-1-butanol, 1-butanol, 2-amino-, r | |
| JCBPETKZIGVZRE-UHFFFAOYNA-N | |
| 2-aminobutan-1-ol | |
| 2723856 | |
| 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 227-476-4
Promotions