missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Acetoxypropionic Acid 98.0+%, TCI America™

Catalog No. p-7118211
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30841G 1 g
A30845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30841G Supplier TCI America Supplier No. A30841G

Chemical Identifiers

CAS 18668-00-3
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.12
MDL Number MFCD16036178
InChI Key WTLNOANVTIKPEE-UHFFFAOYNA-N
Synonym (+)-O-Acetyl-D-lactic Acid
PubChem CID 5325906
IUPAC Name 2-(acetyloxy)propanoic acid
SMILES CC(OC(C)=O)C(O)=O

Specifications

Color Yellow
Boiling Point 82°C
Quantity 1 g
Formula Weight 132.12
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-(+)-2-Acetoxypropionic Acid
Recommended Storage Refrigerator