missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2-Acetoxypropionic Acid 98.0+%, TCI America™

Catalog No. a30841g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
A30841G 1 g
A30845G 5 g
2 options

Catalog No. A30841G

Supplier: TCI America A30841G

Only null left

Chemical Identifiers

CAS 18668-00-3
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.12
MDL Number MFCD16036178
InChI Key WTLNOANVTIKPEE-UHFFFAOYNA-N
Synonym (+)-O-Acetyl-D-lactic Acid
PubChem CID 5325906
IUPAC Name 2-(acetyloxy)propanoic acid
SMILES CC(OC(C)=O)C(O)=O

Specifications

Color Yellow
Boiling Point 82°C
Quantity 1 g
Formula Weight 132.12
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-(+)-2-Acetoxypropionic Acid
Recommended Storage Refrigerator