missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2,3-Diaminopropionic Acid Hydrochloride 98.0+%, TCI America™

Catalog No. p-7126798
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1573100MG 100 mg
D15731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1573100MG Supplier TCI America Supplier No. D1573100MG
Only null left

Chemical Identifiers

CAS 6018-56-0
Molecular Formula C3H9ClN2O2
Molecular Weight (g/mol) 140.567
MDL Number MFCD00070606
InChI Key SKWCZPYWFRTSDD-HSHFZTNMSA-N
Synonym 3-amino-d-alanine hydrochloride, r-2,3-diaminopropanoic acid hydrochloride, h-d-dap-oh hcl, 2r-2,3-diaminopropanoic acid hydrochloride, unii-5452mgq13c, 3-amino-d-alanine monohydrochloride, r---2,3-diaminopropionic acid hydrochloride, d-alanine, 3-amino-, monohydrochloride, d-2,3-diaminopropionic acid hydrochloride, d-2,3-diaminopropionic acid monohydrochloride
PubChem CID 16218517
IUPAC Name (2R)-2,3-diaminopropanoic acid;hydrochloride
SMILES C(C(C(=O)O)N)N.Cl

Specifications

Melting Point 240°C
Color White-Yellow
Quantity 100 mg
Formula Weight 140.57
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-2,3-Diaminopropionic Acid Hydrochloride