missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-2,2-Dimethyl-1,3-dioxolane-4-methanol 97.0+%, TCI America™

Catalog No. D17055G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17055G 5 g
D17051G 1 g
D170525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D17055G Supplier TCI America Supplier No. D17055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14347-78-5
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.159
MDL Number MFCD00003213
InChI Key RNVYQYLELCKWAN-RXMQYKEDSA-N
Synonym r---2,2-dimethyl-1,3-dioxolane-4-methanol, r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, 4r-2,2-dimethyl-1,3-dioxolan-4-yl methanol, r-solketal, r---1,2-o-isopropylideneglycerol, r-2,2-dimethyl-1,3-dioxolane-4-methanol, 2,3-o-isopropylidene-sn-glycerol, d-acetone glycerol, l---1,2-isopropylideneglycerol, r---2,3-o-isopropylideneglycerol
PubChem CID 736056
IUPAC Name [(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]methanol
SMILES CC1(OCC(O1)CO)C

Specifications

Color Yellow
Boiling Point 78°C
Quantity 5 g
Formula Weight 132.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(-)-2,2-Dimethyl-1,3-dioxolane-4-methanol
EINECSNumber (2)-2476&(5)-3554
RTECSNumber JI0400000
TSCA No