missing translation for 'onlineSavingsMsg'
Learn More

(R)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl 98.0+%, TCI America™

Catalog No. p-7122804
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32021G 1 g
B32025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32021G Supplier TCI America Supplier No. B32021G

Chemical Identifiers

CAS 173831-50-0
Molecular Formula C24H22O4
Molecular Weight (g/mol) 374.436
MDL Number MFCD03788935
InChI Key YIAQRNNJNMLGTP-UHFFFAOYSA-N
Synonym r-2,2'-bis methoxymethoxy-1,1'-binaphthalene, s-2,2'-bis methoxymethoxy-1,1'-binaphthalene, r-+-2,2'-bis methoxymethoxy-1,1'-binaphthyl, 2,2'-bis methoxymethoxy-1,1'-binaphthalene, r-2,2'-bis methoxymethoxy-1,1'-binaphthyl, r-+-2,2'-bis methoxymethoxy-1,1'-binaphthalene, 1,1'-binaphthalene, 2,2'-bis methoxymethoxy, s---2,2'-bis methoxymethoxy-1,1'-binaphthalene, s-2,2'-bis methoxymethoxy-1,1'-binaphthyl, r-2,2/'-bis methoxymethoxy-1,1/'-binaphthalene
PubChem CID 10959708
IUPAC Name 2-(methoxymethoxy)-1-[2-(methoxymethoxy)naphthalen-1-yl]naphthalene
SMILES COCOC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)OCOC

Specifications

Melting Point 102°C
Color White-Yellow
Quantity 1 g
Formula Weight 374.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-2,2′-Bis(methoxymethoxy)-1,1′-binaphthyl