missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 102°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 374.44 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (R)-2,2′-Bis(methoxymethoxy)-1,1′-binaphthyl |
Safety and Handling
| TSCA | No |