missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl, 97%, Thermo Scientific Chemicals

$511.95 - $511.95

Chemical Identifiers

CAS 173831-50-0
Molecular Formula C24H22O4
Molecular Weight (g/mol) 374.436
MDL Number MFCD03788935
InChI Key YIAQRNNJNMLGTP-UHFFFAOYSA-N
Synonym r-2,2'-bis methoxymethoxy-1,1'-binaphthalene, s-2,2'-bis methoxymethoxy-1,1'-binaphthalene, r-+-2,2'-bis methoxymethoxy-1,1'-binaphthyl, 2,2'-bis methoxymethoxy-1,1'-binaphthalene, r-2,2'-bis methoxymethoxy-1,1'-binaphthyl, r-+-2,2'-bis methoxymethoxy-1,1'-binaphthalene, 1,1'-binaphthalene, 2,2'-bis methoxymethoxy, s---2,2'-bis methoxymethoxy-1,1'-binaphthalene, s-2,2'-bis methoxymethoxy-1,1'-binaphthyl, r-2,2/'-bis methoxymethoxy-1,1/'-binaphthalene
PubChem CID 10959708
IUPAC Name 2-(methoxymethoxy)-1-[2-(methoxymethoxy)naphthalen-1-yl]naphthalene
SMILES COCOC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)OCOC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3179103
View Documents
Thermo Scientific Chemicals
H3179103
1 g
Each for $511.95
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

173831-50-0
374.436
YIAQRNNJNMLGTP-UHFFFAOYSA-N
10959708
COCOC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)OCOC
C24H22O4
MFCD03788935
r-2,2'-bis methoxymethoxy-1,1'-binaphthalene, s-2,2'-bis methoxymethoxy-1,1'-binaphthalene, r-+-2,2'-bis methoxymethoxy-1,1'-binaphthyl, 2,2'-bis methoxymethoxy-1,1'-binaphthalene, r-2,2'-bis methoxymethoxy-1,1'-binaphthyl, r-+-2,2'-bis methoxymethoxy-1,1'-binaphthalene, 1,1'-binaphthalene, 2,2'-bis methoxymethoxy, s---2,2'-bis methoxymethoxy-1,1'-binaphthalene, s-2,2'-bis methoxymethoxy-1,1'-binaphthyl, r-2,2/'-bis methoxymethoxy-1,1/'-binaphthalene
2-(methoxymethoxy)-1-[2-(methoxymethoxy)naphthalen-1-yl]naphthalene

Specifications

173831-50-0
C24H22O4
1 g
YIAQRNNJNMLGTP-UHFFFAOYSA-N
COCOC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)OCOC
374.436
374.44
(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl
101°C to 105°C
MFCD03788935
r-2,2'-bis methoxymethoxy-1,1'-binaphthalene, s-2,2'-bis methoxymethoxy-1,1'-binaphthalene, r-+-2,2'-bis methoxymethoxy-1,1'-binaphthyl, 2,2'-bis methoxymethoxy-1,1'-binaphthalene, r-2,2'-bis methoxymethoxy-1,1'-binaphthyl, r-+-2,2'-bis methoxymethoxy-1,1'-binaphthalene, 1,1'-binaphthalene, 2,2'-bis methoxymethoxy, s---2,2'-bis methoxymethoxy-1,1'-binaphthalene, s-2,2'-bis methoxymethoxy-1,1'-binaphthyl, r-2,2/'-bis methoxymethoxy-1,1/'-binaphthalene
+94.5° (C=1 in THF)
2-(methoxymethoxy)-1-[2-(methoxymethoxy)naphthalen-1-yl]naphthalene
10959708
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

RUO – Research Use Only