missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl, 97%, Thermo Scientific Chemicals

Catalog No. AAH3179103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3179103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3179103 Supplier Thermo Scientific Chemicals Supplier No. H3179103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 173831-50-0
Molecular Formula C24H22O4
Molecular Weight (g/mol) 374.436
MDL Number MFCD03788935
InChI Key YIAQRNNJNMLGTP-UHFFFAOYSA-N
Synonym r-2,2'-bis methoxymethoxy-1,1'-binaphthalene, s-2,2'-bis methoxymethoxy-1,1'-binaphthalene, r-+-2,2'-bis methoxymethoxy-1,1'-binaphthyl, 2,2'-bis methoxymethoxy-1,1'-binaphthalene, r-2,2'-bis methoxymethoxy-1,1'-binaphthyl, r-+-2,2'-bis methoxymethoxy-1,1'-binaphthalene, 1,1'-binaphthalene, 2,2'-bis methoxymethoxy, s---2,2'-bis methoxymethoxy-1,1'-binaphthalene, s-2,2'-bis methoxymethoxy-1,1'-binaphthyl, r-2,2/'-bis methoxymethoxy-1,1/'-binaphthalene
PubChem CID 10959708
IUPAC Name 2-(methoxymethoxy)-1-[2-(methoxymethoxy)naphthalen-1-yl]naphthalene
SMILES COCOC1=C(C2=CC=CC=C2C=C1)C3=C(C=CC4=CC=CC=C43)OCOC

Specifications

Melting Point 101°C to 105°C
Quantity 1 g
Optical Rotation +94.5° (C=1 in THF)
Formula Weight 374.44
Percent Purity 97%
Chemical Name or Material (R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl

RUO – Research Use Only