missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-1-(tert-Butoxycarbonyl)-3-pyrrolidinol 98.0+%, TCI America™
$67.11 - $201.32
Chemical Identifiers
| CAS | 109431-87-0 |
|---|---|
| Molecular Formula | C9H17NO3 |
| Molecular Weight (g/mol) | 187.24 |
| MDL Number | MFCD01317838 |
| InChI Key | APCBTRDHCDOPNY-UHFFFAOYNA-N |
| Synonym | r-1-boc-3-hydroxypyrrolidine, r-1-n-boc-3-hydroxypyrrolidine, r---n-boc-3-pyrrolidinol, r-n-tert-butoxycarbonyl-3-hydroxypyrrolidine, tert-butyl 3r-3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-pyrrolidinol, r-bochp, r-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-hydropyrrolidine, r---boc-3-pyrrolidinol |
| PubChem CID | 6544479 |
| IUPAC Name | tert-butyl 3-hydroxypyrrolidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CCC(O)C1 |
Chemical Identifiers
| 109431-87-0 | |
| 187.24 | |
| APCBTRDHCDOPNY-UHFFFAOYNA-N | |
| 6544479 | |
| CC(C)(C)OC(=O)N1CCC(O)C1 |
| C9H17NO3 | |
| MFCD01317838 | |
| r-1-boc-3-hydroxypyrrolidine, r-1-n-boc-3-hydroxypyrrolidine, r---n-boc-3-pyrrolidinol, r-n-tert-butoxycarbonyl-3-hydroxypyrrolidine, tert-butyl 3r-3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-pyrrolidinol, r-bochp, r-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-hydropyrrolidine, r---boc-3-pyrrolidinol | |
| tert-butyl 3-hydroxypyrrolidine-1-carboxylate |
Specifications
| 109431-87-0 | |
| White-Yellow | |
| MFCD01317838 | |
| r-1-boc-3-hydroxypyrrolidine, r-1-n-boc-3-hydroxypyrrolidine, r---n-boc-3-pyrrolidinol, r-n-tert-butoxycarbonyl-3-hydroxypyrrolidine, tert-butyl 3r-3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-pyrrolidinol, r-bochp, r-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-hydropyrrolidine, r---boc-3-pyrrolidinol | |
| CC(C)(C)OC(=O)N1CCC(O)C1 | |
| 187.24 | |
| 187.24 | |
| Crystalline Powder |
| 61°C | |
| C9H17NO3 | |
| 1 g | |
| APCBTRDHCDOPNY-UHFFFAOYNA-N | |
| tert-butyl 3-hydroxypyrrolidine-1-carboxylate | |
| 6544479 | |
| ≥98.0% (GC) | |
| (R)-1-(tert-Butoxycarbonyl)-3-pyrrolidinol |
Safety and Handling
TSCA : No