missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(tert-Butoxycarbonyl)-3-pyrrolidinol 98.0+%, TCI America™

Catalog No. B30541G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30541G 1 g
B30545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. B30541G missing translation for 'mfr' TCI America missing translation for 'supplierNo' B30541G

Chemical Identifiers

CAS 109431-87-0
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.24
MDL Number MFCD01317838
InChI Key APCBTRDHCDOPNY-UHFFFAOYNA-N
Synonym r-1-boc-3-hydroxypyrrolidine, r-1-n-boc-3-hydroxypyrrolidine, r---n-boc-3-pyrrolidinol, r-n-tert-butoxycarbonyl-3-hydroxypyrrolidine, tert-butyl 3r-3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-pyrrolidinol, r-bochp, r-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-hydropyrrolidine, r---boc-3-pyrrolidinol
PubChem CID 6544479
IUPAC Name tert-butyl 3-hydroxypyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(O)C1

Specifications

Melting Point 61°C
Color White-Yellow
Quantity 1 g
Formula Weight 187.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-1-(tert-Butoxycarbonyl)-3-pyrrolidinol