missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(tert-Butoxycarbonyl)-3-hydroxypiperidine 98.0+%, TCI America™

Catalog No. p-7123405
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41991G 1 g
B41995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41991G Supplier TCI America Supplier No. B41991G
Only null left

Chemical Identifiers

CAS 143900-43-0
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD04115306
InChI Key UIJXHKXIOCDSEB-MRVPVSSYSA-N
Synonym r-1-boc-3-hydroxypiperidine, r-tert-butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl 3r-3-hydroxypiperidine-1-carboxylate, r-1-n-boc-3-hydroxypiperidine, r-n-boc-3-hydroxypiperidine, r-1-boc-3-hydroxylpiperidine, 3r-1-tert-butoxycarbonyl-3-hydroxypiperidine, 1-piperidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 3r, r-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, r-3-hydroxy-1-tert-butoxycarbonyl piperidine
PubChem CID 1514398
IUPAC Name tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)O

Specifications

Melting Point 50°C
Color White
Quantity 1 g
Formula Weight 201.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material (R)-1-(tert-Butoxycarbonyl)-3-hydroxypiperidine