missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol 98.0+%, TCI America™
$913.38 - $3502.56
Chemical Identifiers
| CAS | 161511-90-6 |
|---|---|
| Molecular Formula | C9H17NO3 |
| Molecular Weight (g/mol) | 187.239 |
| MDL Number | MFCD03790937 |
| InChI Key | XIRUXUKRGUFEKC-SSDOTTSWSA-N |
| Synonym | (R)-1-Boc-2-azetidinemethanol |
| PubChem CID | 11074142 |
| IUPAC Name | tert-butyl (2R)-2-(hydroxymethyl)azetidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CCC1CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B2174100MG
|
TCI America
B2174100MG |
100 mg |
Each for $913.38
|
|
|||||
|
B21741G
|
TCI America
B21741G |
1 g |
Each for $3,502.56
|
|
|||||
Chemical Identifiers
| 161511-90-6 | |
| 187.239 | |
| XIRUXUKRGUFEKC-SSDOTTSWSA-N | |
| 11074142 | |
| CC(C)(C)OC(=O)N1CCC1CO |
| C9H17NO3 | |
| MFCD03790937 | |
| (R)-1-Boc-2-azetidinemethanol | |
| tert-butyl (2R)-2-(hydroxymethyl)azetidine-1-carboxylate |
Specifications
| 161511-90-6 | |
| C9H17NO3 | |
| (R)-1-Boc-2-azetidinemethanol | |
| CC(C)(C)OC(=O)N1CCC1CO | |
| 100 mg | |
| 11074142 | |
| ≥98.0% (GC) | |
| (R)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol |
| Yellow | |
| MFCD03790937 | |
| XIRUXUKRGUFEKC-SSDOTTSWSA-N | |
| tert-butyl (2R)-2-(hydroxymethyl)azetidine-1-carboxylate | |
| 187.239 | |
| 187.24 | |
| Liquid |
Safety and Handling
TSCA : No