missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol 98.0+%, TCI America™

Catalog No. B2174100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2174100MG 100 mg
B21741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2174100MG Supplier TCI America Supplier No. B2174100MG
Only null left

Chemical Identifiers

CAS 161511-90-6
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.239
MDL Number MFCD03790937
InChI Key XIRUXUKRGUFEKC-SSDOTTSWSA-N
Synonym (R)-1-Boc-2-azetidinemethanol
PubChem CID 11074142
IUPAC Name tert-butyl (2R)-2-(hydroxymethyl)azetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC1CO

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 187.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-1-(tert-Butoxycarbonyl)-2-azetidinemethanol