missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Phenylethylamine 99.0+%, TCI America™

Catalog No. P079425ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P079425ML 25 mL
P0794100ML 100 mL
P0794500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P079425ML Supplier TCI America Supplier No. P079425ML

Chemical Identifiers

CAS 3886-69-9
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00064405
InChI Key RQEUFEKYXDPUSK-SSDOTTSWSA-N
Synonym r-1-phenylethanamine, r-+-1-phenylethylamine, 1r-1-phenylethanamine, r-1-phenylethylamine, d-alpha-methylbenzylamine, 1r-1-phenylethan-1-amine, r-+-alpha-methylbenzylamine, r-alpha-methylbenzenemethanamine, unii-v022zk8gz5, r-alfa-methylbenzylamine
PubChem CID 643189
ChEBI CHEBI:35322
IUPAC Name (1R)-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N

Specifications

Color Yellow
Boiling Point 85°C
Quantity 25 mL
UN Number 2922
Formula Weight 121.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-1-Phenylethylamine
EINECSNumber (3)-2692
RTECSNumber DP5775000
TSCA Yes