missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Phenylethylamine, 99+%, ee 99+%

Catalog No. p-7047400
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1911706 5 g
AAL1911714 25 g
AAL1911722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1911706 Supplier Thermo Scientific Chemicals Supplier No. L1911706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3886-69-9 | C8H11N | 121.183 g/mol

(R)-(+)-1-Phenylethylamine is used as an intermediate in organic synthesis. It is also used to determine the enantiomeric purity of acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1-Phenylethylamine is used as an intermediate in organic synthesis. It is also used to determine the enantiomeric purity of acids.

Solubility
Miscible with water and chloroform.

Notes
Store in a cool place. Air Sensitive. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.

Chemical Identifiers

CAS 3886-69-9
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00064405
InChI Key RQEUFEKYXDPUSK-SSDOTTSWSA-N
Synonym r-1-phenylethanamine, r-+-1-phenylethylamine, 1r-1-phenylethanamine, r-1-phenylethylamine, d-alpha-methylbenzylamine, 1r-1-phenylethan-1-amine, r-+-alpha-methylbenzylamine, r-alpha-methylbenzenemethanamine, unii-v022zk8gz5, r-alfa-methylbenzylamine
PubChem CID 643189
ChEBI CHEBI:35322
IUPAC Name (1R)-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)N

Specifications

Melting Point -10°C
Density 0.948
Boiling Point 187°C
Flash Point 71°C (159°F)
Odor Amine-like
Refractive Index 1.526
Quantity 5 g
UN Number UN2735
Beilstein 2410916
Sensitivity Air Sensitive
Merck Index 14,6026
Solubility Information Miscible with water and chloroform.
Optical Rotation +30° (c=10 in Ethanol)
Formula Weight 121.18
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (R)-(+)-1-Phenylethylamine
Show More Show Less
Hazard Category H227-H302+H312-H314
Hazard Statement GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 223-423-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only