missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Phenylethyl isocyanate, 99%

Catalog No. AAL1030506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1030506 5 g
AAL1030503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1030506 Supplier Thermo Scientific Chemicals Supplier No. L1030506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33375-06-3 | C9H9NO | 147.177 g/mol

(R)-1-phenylethyl isocyanate is used in the derivatisation of alcohols, thiocarbamates and hydroxy fatty acids. (R)-1-phenylethyl isocyanate moderately inhibited both BP-DNA adducts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-1-phenylethyl isocyanate is used in the derivatisation of alcohols, thiocarbamates and hydroxy fatty acids. (R)-1-phenylethyl isocyanate moderately inhibited both BP-DNA adducts.

Solubility
Decomposes in water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture. Store at 4°C.

Chemical Identifiers

CAS 33375-06-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00063015
InChI Key JJSCUXAFAJEQGB-MRVPVSSYSA-N
Synonym r-+-1-phenylethyl isocyanate, 1r-1-isocyanatoethyl benzene, r-+-alpha-methylbenzyl isocyanate, benzene, 1r-1-isocyanatoethyl, isocyanic acid r-+-1-phenylethyl ester, isocyanic acid r-+-alpha-methylbenzyl ester, isocyanic acid-r-+-methylbenzyl ester, 1-isocyanatoethyl benzene #, +-alpha-methylbenzyl isocyanate, pubchem8096
PubChem CID 7018262
IUPAC Name [(1R)-1-isocyanatoethyl]benzene
SMILES CC(C1=CC=CC=C1)N=C=O

Specifications

Density 1.045
Boiling Point 55°C to 56°C (2.5 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.5124
Quantity 5 g
UN Number UN2206
Beilstein 3196930
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Optical Rotation +10.5° (Neat)
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material (R)-(+)-1-Phenylethyl isocyanate
Show More Show Less
Hazard Category H227-H315-H319-H330-H334-H335
Hazard Statement GHS H Statement
H331-H334-H227-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P284-P285-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 251-485-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only