missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Phenylethanol, 99%, ee 97+%

Catalog No. AAL1929606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1929606 5 g
AAL1929614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1929606 Supplier Thermo Scientific Chemicals Supplier No. L1929606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1517-69-7 | C8H10O | 122.17 g/mol

(R)-(+)-1-Phenylethanol is used in the synthesis of optically active products. It is also useful in the determination of enantiomeric purity and for resolutions of acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1-Phenylethanol is used in the synthesis of optically active products. It is also useful in the determination of enantiomeric purity and for resolutions of acids.

Solubility
Miscible with organic solvents and slightly miscible with water.

Notes
Incompatible with strong acids and strong oxidizing agents.

Chemical Identifiers

CAS 1517-69-7
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00064263
InChI Key WAPNOHKVXSQRPX-UHFFFAOYNA-N
Synonym r-1-phenylethanol, r-+-1-phenylethanol, 1r-1-phenylethanol, r-alpha-methylbenzyl alcohol, r-+-sec-phenethyl alcohol, 1r-1-phenylethan-1-ol, r-+-1-phenylethyl alcohol, 1-phenylethanol, r, benzenemethanol, alpha-methyl-, r
PubChem CID 637516
ChEBI CHEBI:45616
SMILES CC(O)C1=CC=CC=C1

Specifications

Melting Point 9°C to 11°C
Density 1.012
Boiling Point 88°C to 89°C (10 mmHg)
Flash Point 85°C (185°F)
Odor Mild
Refractive Index 1.528
Quantity 5 g
UN Number UN2937
Beilstein 2039798
Solubility Information Miscible with organic solvents and slightly miscible with water.
Optical Rotation +45° (c=5 in Methanol)
IUPAC Name 1-phenylethan-1-ol
Formula Weight 122.17
Percent Purity 99%
Assay ee ≥97%
Chemical Name or Material (R)-(+)-1-Phenylethanol
Show More Show Less
Hazard Category H227-H302-H315-H318
Hazard Statement GHS H Statement
H318-H302-H335-H227-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALPHA-METHYLBENZYL ALCOHOL, LIQUID
EINECSNumber 604-814-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only