missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Phenylethane-1,2-diol 98.0+%, TCI America™

Catalog No. P11501G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11501G 1 g
P11505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11501G Supplier TCI America Supplier No. P11501G

Chemical Identifiers

CAS 16355-00-3
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00064262
InChI Key PWMWNFMRSKOCEY-QMMMGPOBSA-N
Synonym r---1-phenyl-1,2-ethanediol, r-1-phenylethane-1,2-diol, 1r-1-phenylethane-1,2-diol, r---1-phenylethane-1,2-diol, r-1-phenyl-1,2-ethanediol, styrene glycol,-, unii-2lzm4b71kf, r-+-1-phenylethane-1,2-diol, 2lzm4b71kf
PubChem CID 2724621
IUPAC Name (1R)-1-phenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(CO)O

Specifications

Melting Point 68°C
Color White
Boiling Point 82°C
Quantity 1 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-1-Phenylethane-1,2-diol