missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-Phenyl-1,3-propanediol, 98%

Catalog No. p-3779453
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC417750010 1 g Glass bottle
AC417750050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC417750010 Supplier Thermo Scientific Chemicals Supplier No. 417750010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103548-16-9 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103548-16-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00145213
InChI Key RRVFYOSEKOTFOG-SECBINFHSA-N
Synonym r-+-1-phenyl-1,3-propanediol, r-1-phenylpropane-1,3-diol, r-1-phenyl-1,3-propanediol, 1r-1-phenylpropane-1,3-diol, r-1-phenyl-1,3-dihydroxypropane, r-3-phenyl-1,3-dihydroxypropane, 1,3-propanediol,1-phenyl-, 1r, r ?-?1-?phenyl-?1,?3-?propanediol, r-+-1-phenyl-1,3-propanediol sum of enantiomers, gc
PubChem CID 2735120
IUPAC Name (1R)-1-phenylpropane-1,3-diol
SMILES C1=CC=C(C=C1)C(CCO)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 62°C to 66°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition +54.50° (22°C c=1,1,2−dichloroeth)
Specific Rotation + 54.5
Formula Weight 152.19
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material (R)-1-Phenyl-1, 3-propanediol
Show More Show Less

RUO – Research Use Only