missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Octyn-3-ol, 98+%, Thermo Scientific Chemicals

Catalog No. p-7047372
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1904503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1904503 Supplier Thermo Scientific Chemicals Supplier No. L1904503
Only null left

(R)-(+)-1-Octyn-3-ol is used as an intermediate for the synthesis of prostaglandins, modified eicosanoids structured with fatty acids attached to a 5 membered ring.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1-Octyn-3-ol is used as an intermediate for the synthesis of prostaglandins, modified eicosanoids structured with fatty acids attached to a 5 membered ring.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 32556-70-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00192370
InChI Key VUGRNZHKYVHZSN-QMMMGPOBSA-N
Synonym r-+-1-octyn-3-ol, 1-octyn-3-ol, 3r, 3r-oct-1-yn-3-ol, r-1-octyn-3-ol, unii-az9v2w5sw4, 3r-1-octyn-3-ol, az9v2w5sw4, +-1-octyn-3-ol, 1-octyn-3-ol, r, 1-octyn-3-ol, +
PubChem CID 6999692
IUPAC Name (3R)-oct-1-yn-3-ol
SMILES CCCCCC(C#C)O

Specifications

Density 0.866
Boiling Point 100°C (20 mmHg)
Flash Point 63°C (145°F)
Refractive Index 1.442
Quantity 1 g
Beilstein 2037704
Optical Rotation +9.9° (Neat)
Formula Weight 126.2
Percent Purity ≥98%
Chemical Name or Material (R)-(+)-1-Octyn-3-ol
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only