Learn More
(R)-(+)-1-Octyn-3-ol, 98+%, Thermo Scientific Chemicals
Description
(R)-(+)-1-Octyn-3-ol is used as an intermediate for the synthesis of prostaglandins, modified eicosanoids structured with fatty acids attached to a 5 membered ring.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(R)-(+)-1-Octyn-3-ol is used as an intermediate for the synthesis of prostaglandins, modified eicosanoids structured with fatty acids attached to a 5 membered ring.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 0.866 |
| Boiling Point | 100°C (20 mmHg) |
| Flash Point | 63°C (145°F) |
| Refractive Index | 1.442 |
| Quantity | 1 g |
| Beilstein | 2037704 |
| Optical Rotation | +9.9° (Neat) |
| Formula Weight | 126.2 |
| Percent Purity | ≥98% |
| Chemical Name or Material | (R)-(+)-1-Octyn-3-ol |
Safety and Handling
| Hazard Category | H227-H315-H319-H335 |
| Hazard Statement | GHS H Statement H301-H227 Toxic if swallowed. Combustible liquid. |
| Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only