missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-Methyl-3-pyrrolidinol 98.0+%, TCI America™

Catalog No. p-7132922
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19371G 1 g
M19375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19371G Supplier TCI America Supplier No. M19371G
Only null left

Chemical Identifiers

CAS 104641-60-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD03788747
InChI Key FLVFPAIGVBQGET-UHFFFAOYNA-N
Synonym 3r-1-methylpyrrolidin-3-ol, r---1-methyl-3-pyrrolidinol, r-3-hydroxy-1-methyl-pyrrolidine, r-1-methyl-3-pyrrolidinol, r-1-methylpyrrolidin-3-ol, r-3-hydroxy-1-methylpyrrolidine, r---1-methyl-3-hydroxypyrrolidine, 3-pyrrolidinol, 1-methyl-, 3r, r-1-methyl-pyrrolidin-3-ol, unii-6hvw4ew50x
PubChem CID 6951332
IUPAC Name 1-methylpyrrolidin-3-ol
SMILES CN1CCC(O)C1

Specifications

Color Yellow
Boiling Point 106°C
Quantity 1 g
Formula Weight 101.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-1-Methyl-3-pyrrolidinol