missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Methyl-3-hydroxypyrrolidine, 99%

Catalog No. AAL1969403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1969403 1 g
AAL1969406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1969403 Supplier Thermo Scientific Chemicals Supplier No. L1969403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 104641-60-3 | C5H11NO | 101.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104641-60-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD03788747
InChI Key FLVFPAIGVBQGET-UHFFFAOYNA-N
Synonym 3r-1-methylpyrrolidin-3-ol, r---1-methyl-3-pyrrolidinol, r-3-hydroxy-1-methyl-pyrrolidine, r-1-methyl-3-pyrrolidinol, r-1-methylpyrrolidin-3-ol, r-3-hydroxy-1-methylpyrrolidine, r---1-methyl-3-hydroxypyrrolidine, 3-pyrrolidinol, 1-methyl-, 3r, r-1-methyl-pyrrolidin-3-ol, unii-6hvw4ew50x
PubChem CID 6951332
SMILES CN1CCC(O)C1

Specifications

Density 0.92
Boiling Point 103°C to 106°C (35 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.464
Quantity 1 g
Beilstein 4349393
Sensitivity Air sensitive; Hygroscopic
IUPAC Name 1-methylpyrrolidin-3-ol
Formula Weight 101.15
Percent Purity 99%
Assay ee 99%
Chemical Name or Material (R)-(-)-1-Methyl-3-hydroxypyrrolidine
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only