missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-BOC-3-methylpiperazine, 99%

Catalog No. AC432050010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432050010 1 g Glass bottle
AC432050050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432050010 Supplier Thermo Scientific Chemicals Supplier No. 432050010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 163765-44-4 | C10H20N2O2 | 200.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 163765-44-4
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
MDL Number MFCD02683205
InChI Key FMLPQHJYUZTHQS-MRVPVSSYSA-N
Synonym r-1-boc-3-methylpiperazine, r-4-boc-2-methylpiperazine, r-4-n-boc-2-methylpiperazine, r-tert-butyl 3-methylpiperazine-1-carboxylate, tert-butyl 3r-3-methylpiperazine-1-carboxylate, r-4-n-boc-2-methyl-piperazine, r-1-boc-3-methyl-piperazine, r-4bocmp, r-n4-boc-2-methylpiperazine, 3r-1-boc-3-methylpiperazine
PubChem CID 2756811
IUPAC Name tert-butyl (3R)-3-methylpiperazine-1-carboxylate
SMILES CC1CN(CCN1)C(=O)OC(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 44.4°C to 46.6°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation + 15
Formula Weight 200.28
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material (R)-1-BOC-3-Methylpiperazine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only