missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-Boc-3-methylpiperazine, 97%

Catalog No. AAH2735003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2735003 1 g
AAH2735006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2735003 Supplier Thermo Scientific Chemicals Supplier No. H2735003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 163765-44-4 | C10H20N2O2 | 200.282 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 163765-44-4
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD02683205
InChI Key FMLPQHJYUZTHQS-MRVPVSSYSA-N
Synonym r-1-boc-3-methylpiperazine, r-4-boc-2-methylpiperazine, r-4-n-boc-2-methylpiperazine, r-tert-butyl 3-methylpiperazine-1-carboxylate, tert-butyl 3r-3-methylpiperazine-1-carboxylate, r-4-n-boc-2-methyl-piperazine, r-1-boc-3-methyl-piperazine, r-4bocmp, r-n4-boc-2-methylpiperazine, 3r-1-boc-3-methylpiperazine
PubChem CID 2756811
IUPAC Name tert-butyl (3R)-3-methylpiperazine-1-carboxylate
SMILES CC1CN(CCN1)C(=O)OC(C)(C)C

Specifications

Melting Point 40°C to 45°C
Quantity 1 g
Optical Rotation +16° (c=1 in Dioxane)
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material (R)-(+)-1-Boc-3-methylpiperazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only