missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Boc-3-hydroxypyrrolidine, 98%

Catalog No. AAH2759003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2759003 1 g
AAH2759006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2759003 Supplier Thermo Scientific Chemicals Supplier No. H2759003

CAS: 109431-87-0 | C9H17NO3 | 187.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109431-87-0
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.24
MDL Number MFCD01317838
InChI Key APCBTRDHCDOPNY-UHFFFAOYNA-N
Synonym r-1-boc-3-hydroxypyrrolidine, r-1-n-boc-3-hydroxypyrrolidine, r---n-boc-3-pyrrolidinol, r-n-tert-butoxycarbonyl-3-hydroxypyrrolidine, tert-butyl 3r-3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-pyrrolidinol, r-bochp, r-tert-butyl 3-hydroxypyrrolidine-1-carboxylate, r-n-boc-3-hydropyrrolidine, r---boc-3-pyrrolidinol
PubChem CID 6544479
IUPAC Name tert-butyl (3R)-3-hydroxypyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(O)C1

Specifications

Melting Point 62°C to 65°C
Quantity 1 g
Formula Weight 187.24
Percent Purity 98%
Chemical Name or Material (R)-(-)-1-Boc-3-hydroxypyrrolidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only