missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-Boc-3-hydroxypiperidine, 97%

Catalog No. AAH5255603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5255603 1 g
AAH5255606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5255603 Supplier Thermo Scientific Chemicals Supplier No. H5255603

CAS: 143900-43-0 | C10H19NO3 | 201.266 g/mol

(R)-1-Boc-3-hydroxypiperidine has been used as a reactant for the synthesis of piperidinyl and pyrrolidinyl butyrates. It has also been used for the synthesis of constrained (-)-S-adenosyl-L-homocysteine (SAH) analogs as DNA methyltransferase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-1-Boc-3-hydroxypiperidine has been used as a reactant for the synthesis of piperidinyl and pyrrolidinyl butyrates. It has also been used for the synthesis of constrained (-)-S-adenosyl-L-homocysteine (SAH) analogs as DNA methyltransferase inhibitors.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 143900-43-0
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD04115306
InChI Key UIJXHKXIOCDSEB-MRVPVSSYSA-N
Synonym r-1-boc-3-hydroxypiperidine, r-tert-butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl 3r-3-hydroxypiperidine-1-carboxylate, r-1-n-boc-3-hydroxypiperidine, r-n-boc-3-hydroxypiperidine, r-1-boc-3-hydroxylpiperidine, 3r-1-tert-butoxycarbonyl-3-hydroxypiperidine, 1-piperidinecarboxylic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 3r, r-3-hydroxy-piperidine-1-carboxylic acid tert-butyl ester, r-3-hydroxy-1-tert-butoxycarbonyl piperidine
PubChem CID 1514398
IUPAC Name tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)O

Specifications

Melting Point 46°C to 50°C
Flash Point 104°C (219°F)
Quantity 1 g
Optical Rotation −23° (c=5 in Methanol)
Formula Weight 201.27
Percent Purity 97%
Chemical Name or Material (R)-1-Boc-3-hydroxypiperidine
Hazard Category H301-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only