missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Boc-3-(hydroxymethyl)piperidine, 97%

Catalog No. AAH5255903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5255903 1 g
AAH5255906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5255903 Supplier Thermo Scientific Chemicals Supplier No. H5255903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 140695-85-8 | C11H21NO3 | 215.293 g/mol

(R)-(-)-1-Boc-3-(hydroxymethyl)piperidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-1-Boc-3-(hydroxymethyl)piperidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 140695-85-8
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.293
MDL Number MFCD02683202
InChI Key OJCLHERKFHHUTB-SECBINFHSA-N
Synonym r-1-boc-3-hydroxymethyl piperidine, r-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, r-1-boc-3-hyroxymethyl piperidine, tert-butyl 3r-3-hydroxymethyl piperidine-1-carboxylate, r-n-boc-3-hydroxymethyl piperidine, r-n-boc-3-piperidinemethanol, r-1-n-boc-3-hydroxymethyl piperidine, r-1-n-boc-3-hydroxymethyl-piperidine, r---1-boc-3-hydroxymethyl piperidine
PubChem CID 1514447
IUPAC Name tert-butyl (3R)-3-(hydroxymethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)CO

Specifications

Melting Point 91°C to 94°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Optical Rotation −20° (c=2 in Methanol)
Formula Weight 215.29
Percent Purity 97%
Chemical Name or Material (R)-(-)-1-Boc-3-(hydroxymethyl)piperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only