missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Boc-3-bromopyrrolidine, 95%

Catalog No. p-7038303
Change view
Click to view available options
Quantity:
1 g
250 mg
Catalog No. Quantity
AAH57228MD 250 mg
AAH5722803 1 g
2 options

Catalog No. AAH57228MD

Supplier: Thermo Scientific Chemicals H57228MD

Only null left

CAS: 569660-97-5 | C9H16BrNO2 | 250.136 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Chemical Identifiers

CAS 569660-97-5
Molecular Formula C9H16BrNO2
Molecular Weight (g/mol) 250.136
MDL Number MFCD17214727
InChI Key QJTKPXFJOXKUEY-SSDOTTSWSA-N
Synonym r-tert-butyl 3-bromopyrrolidine-1-carboxylate, r-3-bromo-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 3r-3-bromopyrrolidine-1-carboxylate, r---1-boc-3-bromopyrrolidine, s-+-1-boc-3-bromopyrrolidine, 1-boc-r-3-bromopyrrolidine, tert-butyl r-3-bromopyrrolidine-1-carboxylate, r-3-bromo-pyrrolidine-1-carboxylic acid tert-but, r-3-bromopyrrolidine-1-carboxylic acid tert-butyl ester
PubChem CID 34179476
IUPAC Name tert-butyl (3R)-3-bromopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)Br

Specifications

Quantity 250 mg
UN Number UN3077
Optical Rotation −33° (c=0.8 in Chloroform)
Formula Weight 250.14
Percent Purity 95%
Chemical Name or Material (R)-(-)-1-Boc-3-bromopyrrolidine
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only