Learn More
(R)-1-BOC-3-(Aminomethyl)piperidine, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 140645-23-4 |
|---|---|
| Molecular Formula | C11H22N2O2 |
| Molecular Weight (g/mol) | 214.31 |
| InChI Key | WPWXYQIMXTUMJB-SECBINFHSA-N |
| Synonym | r-1-boc-3-aminomethyl piperidine, r-n-boc-3-aminomethylpiperidine, r-tert-butyl 3-aminomethyl piperidine-1-carboxylate, r-1-n-boc-3-aminomethyl piperidine, tert-butyl 3r-3-aminomethyl piperidine-1-carboxylate, r-3-aminomethyl-1-n-boc-piperidine, r-3-aminomethyl-1-boc-piperidine, r-3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, 3r-3-aminomethyl-n-tert-butoxycarbonylpiperidine |
| PubChem CID | 1502023 |
| IUPAC Name | tert-butyl (3R)-3-(aminomethyl)piperidine-1-carboxylate |
| SMILES | CC(C)(C)OC(=O)N1CCCC(C1)CN |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC436420010
|
Thermo Scientific Chemicals
436420010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC436420050
|
Thermo Scientific Chemicals
436420050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 140645-23-4 | |
| 214.31 | |
| r-1-boc-3-aminomethyl piperidine, r-n-boc-3-aminomethylpiperidine, r-tert-butyl 3-aminomethyl piperidine-1-carboxylate, r-1-n-boc-3-aminomethyl piperidine, tert-butyl 3r-3-aminomethyl piperidine-1-carboxylate, r-3-aminomethyl-1-n-boc-piperidine, r-3-aminomethyl-1-boc-piperidine, r-3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, 3r-3-aminomethyl-n-tert-butoxycarbonylpiperidine | |
| tert-butyl (3R)-3-(aminomethyl)piperidine-1-carboxylate |
| C11H22N2O2 | |
| WPWXYQIMXTUMJB-SECBINFHSA-N | |
| 1502023 | |
| CC(C)(C)OC(=O)N1CCCC(C1)CN |
Specifications
| 140645-23-4 | |
| 100.0 | |
| 96% min. (GC) | |
| C11H22N2O2 | |
| r-1-boc-3-aminomethyl piperidine, r-n-boc-3-aminomethylpiperidine, r-tert-butyl 3-aminomethyl piperidine-1-carboxylate, r-1-n-boc-3-aminomethyl piperidine, tert-butyl 3r-3-aminomethyl piperidine-1-carboxylate, r-3-aminomethyl-1-n-boc-piperidine, r-3-aminomethyl-1-boc-piperidine, r-3-aminomethyl-piperidine-1-carboxylic acid tert-butyl ester, 3r-3-aminomethyl-n-tert-butoxycarbonylpiperidine | |
| CC(C)(C)OC(=O)N1CCCC(C1)CN | |
| 214.31 | |
| 214.31 | |
| (R)-1-BOC-3-(Aminomethyl)piperidine |
| 96.0 | |
| Authentic | |
| Glass bottle | |
| 1g | |
| WPWXYQIMXTUMJB-SECBINFHSA-N | |
| tert-butyl (3R)-3-(aminomethyl)piperidine-1-carboxylate | |
| 1502023 | |
| 97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
RUO â Research Use Only