missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Amino-2-propanol 98.0+%, TCI America™

Catalog No. A09745G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09745G 5 g
A09741G 1 g
A097425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A09745G Supplier TCI America Supplier No. A09745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2799-16-8
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00064428
InChI Key HXKKHQJGJAFBHI-GSVOUGTGSA-N
Synonym r---1-amino-2-propanol, r-1-aminopropan-2-ol, r-1-amino-2-propanol, 2r-1-aminopropan-2-ol, r---isopropanolamine, 2-propanol, 1-amino-, 2r, 2r---hydroxypropylamine, r---1-aminopropan-2-ol, 2r---2-hydroxypropylamine
PubChem CID 439938
ChEBI CHEBI:15675
IUPAC Name (2R)-1-aminopropan-2-ol
SMILES C[C@@H](O)CN

Specifications

Color White-Yellow
Boiling Point 160°C
Quantity 5 g
UN Number 3259
Formula Weight 75.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material (R)-(-)-1-Amino-2-propanol