missing translation for 'onlineSavingsMsg'
Learn More

(R)-1,4-Diazabicyclo[4.3.0]nonane, 97%

Catalog No. AAH52554MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52554MD 250 mg
AAH5255403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52554MD Supplier Thermo Scientific Chemicals Supplier No. H52554MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 96193-27-0 | C7H14N2 | 126.203 g/mol

(R)-1,4-Diazabicyclo[4.3.0]nonane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-1,4-Diazabicyclo[4.3.0]nonane is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions.

Chemical Identifiers

CAS 96193-27-0
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.203
MDL Number MFCD09752820
InChI Key FTTATHOUSOIFOQ-SSDOTTSWSA-N
Synonym r-octahydropyrrolo 1,2-a pyrazine, r-1,4-diazabicyclo 4.3.0 nonane, 8ar-octahydropyrrolo 1,2-a pyrazine, r-octahydro-pyrrolo 1,2-a pyrazine, 8ar-octahydropyrrolo 1,2-a piperazine, r-1,4-diazobicyclo 4.3.0 nonane, 6r-1,4-diazabicyclo 4.3.0 nonane, pyrrolo 1,2-a pyrazine, octahydro-, 8ar-9ci, pyrrolo 1,2-a pyrazine, octahydro-, r, pyrrolo 1,2-a pyrazine, octahydro-, 8ar
PubChem CID 7004239
IUPAC Name (8aR)-1,2,3,4,6,7,8,8a-octahydropyrrolo[1,2-a]pyrazine
SMILES C1CC2CNCCN2C1

Specifications

Boiling Point 65°C to 69°C (3 mmHg)
Quantity 250 mg
UN Number UN3267
Solubility Information Slightly soluble in water.
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material (R)-1,4-Diazabicyclo[4.3.0]nonane
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only