missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(4-Chlorophenyl)ethylamine, 97%, 98% ee, Thermo Scientific™

Catalog No. AC370650010
Change view
Click to view available options
Glass bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC370650010 Glass bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC370650010 Supplier Thermo Scientific Chemicals Supplier No. 370650010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 27298-99-3
Density 1.1150g/mL
Boiling Point 232.0°C
Flash Point 115°C
Enantiomeric Excess 90.0% min.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H10ClN
Refractive Index 1.5410 to 1.5430
Linear Formula ClC6H4CH(CH3)NH2
Quantity 1g
MDL Number MFCD00671639
Specific Gravity 1.115
Specific Rotation Condition +23.5° (20°C c=2,methanol)
Synonym r-1-4-chlorophenyl ethylamine, r-1-4-chlorophenyl ethanamine, 1r-1-4-chlorophenyl ethanamine, r-+-1-4-chlorophenyl ethylamine, 1r-1-4-chlorophenyl ethan-1-amine, benzenemethanamine, 4-chloro-alpha-methyl-, alphar, pubchem15222, r-4-chloro-, a-methylbenzylamine, 1r-1-4-chlorophenyl ethylamine, r-1-4-chloro-phenyl ethylamine
Specific Rotation + 23.50
Solubility Information Solubility in water: moderately soluble
InChI Key PINPOEWMCLFRRB-ZCFIWIBFSA-N
SMILES CC(C1=CC=C(C=C1)Cl)N
IUPAC Name (1R)-1-(4-chlorophenyl)ethanamine
Molecular Weight (g/mol) 155.63
PubChem CID 1715226
Formula Weight 155.63
Percent Purity 97%,98% ee
Chemical Name or Material (R)-1-(4-Chlorophenyl)ethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

RUO – Research Use Only