missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1,2-Propanediol 97.0+%, TCI America™

Catalog No. p-7134977
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11525G 5 g
P115225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11525G Supplier TCI America Supplier No. P11525G
Only null left

Chemical Identifiers

CAS 4254-14-2
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00066248
InChI Key DNIAPMSPPWPWGF-GSVOUGTGSA-N
Synonym r---1,2-propanediol, r-propane-1,2-diol, 2r-propane-1,2-diol, r-1,2-propanediol, r-propylene glycol, 2r-1,2-propanediol, r---1,2-dihydroxypropane, unii-602hn5l69h
PubChem CID 259994
ChEBI CHEBI:28972
IUPAC Name (2R)-propane-1,2-diol
SMILES CC(CO)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 76.10
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(-)-1,2-Propanediol
EINECSNumber (2)-0234
TSCA No