missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1,2-Propanediol, 98%

Catalog No. p-7026120
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2193703 1 g
AAB2193706 5 g
AAB2193714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2193703 Supplier Thermo Scientific Chemicals Supplier No. B2193703

CAS: 4254-14-2 | C3H8O2 | 76.095 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4254-14-2
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00066248
InChI Key DNIAPMSPPWPWGF-GSVOUGTGSA-N
Synonym r---1,2-propanediol, r-propane-1,2-diol, 2r-propane-1,2-diol, r-1,2-propanediol, r-propylene glycol, 2r-1,2-propanediol, r---1,2-dihydroxypropane, unii-602hn5l69h
PubChem CID 259994
ChEBI CHEBI:28972
IUPAC Name (2R)-propane-1,2-diol
SMILES CC(CO)O

Specifications

Density 1.04
Boiling Point 186°C to 188°C
Flash Point 107°C (224°F)
Refractive Index 1.432
Quantity 1 g
Beilstein 1718872
Sensitivity Hygroscopic
Merck Index 14,7855
Optical Rotation −16.5° (Neat)
Formula Weight 76.1
Percent Purity 98%
Chemical Name or Material (R)-(-)-1,2-Propanediol
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only