missing translation for 'onlineSavingsMsg'
Learn More

(R)-1-(2-Naphthyl)ethylamine 98.0+%, TCI America™

Catalog No. N07241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07241G 1 g
N07245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07241G Supplier TCI America Supplier No. N07241G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3906-16-9
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.243
MDL Number MFCD00085367
InChI Key KHSYYLCXQKCYQX-SECBINFHSA-N
Synonym r-+-1-2-naphthyl ethylamine, r-1-naphthalen-2-yl ethanamine, r-1-2-naphthyl ethylamine, 1r-1-naphthalen-2-yl ethan-1-amine, r-2-1-aminoethyl naphthalene, 1r-1-2-naphthyl ethanamine, 1r-1-naphthalen-2-yl ethanamine, r-2-naphthylethylamine, ksc490g1r, 1r-1-naphth-2-ylethylamine
PubChem CID 1798835
IUPAC Name (1R)-1-naphthalen-2-ylethanamine
SMILES CC(C1=CC2=CC=CC=C2C=C1)N

Specifications

Melting Point 53°C
Color White-Yellow
Boiling Point 166°C
Quantity 1 g
Formula Weight 171.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-1-(2-Naphthyl)ethylamine