missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1,2-Epoxybutane, 98%

CAS: 3760-95-0 | C4H8O | 72.107 g/mol

Supplier:  Thermo Scientific Chemicals H5857906

 View more versions of this product

Catalog No. AAH5857906


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(R)-(+)-1,2-Epoxybutane
0.83
−12°C (10°F)
1.381
5 g
r-+-1,2-epoxybutane, r-2-ethyloxirane, 2r-2-ethyloxirane, r-1,2-epoxybutane, oxirane, ethyl-, 2r, r-epoxybutane, pubchem14098, r-+-ethyloxirane, r-+-butylene oxide
+10° (Neat)
(2R)-2-ethyloxirane
11147670
98%
3760-95-0
60°C to 64°C
C4H8O
MFCD02683444
UN3022
RBACIKXCRWGCBB-SCSAIBSYSA-N
CCC1CO1
72.107
72.11

Chemical Identifiers

3760-95-0
72.107
RBACIKXCRWGCBB-SCSAIBSYSA-N
11147670
CCC1CO1
C4H8O
MFCD02683444
r-+-1,2-epoxybutane, r-2-ethyloxirane, 2r-2-ethyloxirane, r-1,2-epoxybutane, oxirane, ethyl-, 2r, r-epoxybutane, pubchem14098, r-+-ethyloxirane, r-+-butylene oxide
(2R)-2-ethyloxirane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H331-H351-H302-H312-H315-H319-H335
Highly flammable liquid and vapor.
Toxic if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P363-P370+P37

H225-H302+H312-H315-H319-H331-H335-H351

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-BUTYLENE OXIDE, STABILIZED

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only