missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1,2-Epoxybutane, 98%

Catalog No. AAH5857903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5857903 1 g
AAH5857906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5857903 Supplier Thermo Scientific Chemicals Supplier No. H5857903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 3760-95-0 | C4H8O | 72.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3760-95-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD02683444
InChI Key RBACIKXCRWGCBB-SCSAIBSYSA-N
Synonym r-+-1,2-epoxybutane, r-2-ethyloxirane, 2r-2-ethyloxirane, r-1,2-epoxybutane, oxirane, ethyl-, 2r, r-epoxybutane, pubchem14098, r-+-ethyloxirane, r-+-butylene oxide
PubChem CID 11147670
IUPAC Name (2R)-2-ethyloxirane
SMILES CCC1CO1

Specifications

Density 0.83
Boiling Point 60°C to 64°C
Flash Point −12°C (10°F)
Refractive Index 1.381
Quantity 1 g
UN Number UN3022
Optical Rotation +10° (Neat)
Formula Weight 72.11
Percent Purity 98%
Chemical Name or Material (R)-(+)-1,2-Epoxybutane
Hazard Category H225-H302+H312-H315-H319-H331-H335-H351
Hazard Statement GHS H Statement
H225-H331-H351-H302-H312-H315-H319-H335
Highly flammable liquid and vapor.
Toxic if inhaled.
Suspected of causing cancer.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P332+P313-P363-P370+P37
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-BUTYLENE OXIDE, STABILIZED
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only