missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-1,2-Butanediol 98.0+%, TCI America™
$55.92 - $169.63
Chemical Identifiers
| CAS | 40348-66-1 |
|---|---|
| Molecular Formula | C4H10O2 |
| Molecular Weight (g/mol) | 90.12 |
| MDL Number | MFCD09953765 |
| InChI Key | BMRWNKZVCUKKSR-UHFFFAOYNA-N |
| Synonym | (R)-1,2-Butylene Glycol, (R)-1,2-Dihydroxybutane |
| PubChem CID | 641012 |
| ChEBI | CHEBI:52685 |
| IUPAC Name | butane-1,2-diol |
| SMILES | CCC(O)CO |
Chemical Identifiers
| 40348-66-1 | |
| 90.12 | |
| BMRWNKZVCUKKSR-UHFFFAOYNA-N | |
| 641012 | |
| butane-1,2-diol |
| C4H10O2 | |
| MFCD09953765 | |
| (R)-1,2-Butylene Glycol, (R)-1,2-Dihydroxybutane | |
| CHEBI:52685 | |
| CCC(O)CO |
Specifications
| 40348-66-1 | |
| 60°C | |
| MFCD09953765 | |
| (R)-1,2-Butylene Glycol, (R)-1,2-Dihydroxybutane | |
| CCC(O)CO | |
| 90.12 | |
| CHEBI:52685 | |
| ≥98.0% (GC) | |
| (R)-1,2-Butanediol |
| Yellow | |
| C4H10O2 | |
| 1 g | |
| BMRWNKZVCUKKSR-UHFFFAOYNA-N | |
| butane-1,2-diol | |
| 641012 | |
| 90.12 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0235
TSCA : No