missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-(1-Naphthyl)ethylamine 99.0+%, TCI America™

Catalog No. N04821G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04821G 1 g
N04825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04821G Supplier TCI America Supplier No. N04821G

Chemical Identifiers

CAS 3886-70-2
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.243
MDL Number MFCD00064114
InChI Key RTCUCQWIICFPOD-SECBINFHSA-N
Synonym r-+-1-1-naphthyl ethylamine, r-1-naphthalen-1-yl ethanamine, 1r-1-naphthalen-1-yl ethan-1-amine, r-1-1-naphthyl ethylamine, 1r-1-naphthalen-1-ylethanamine, 1r-1-naphthylethylamine, r-+-alpha-1-naphthyl ethylamine, 1r-1-1-naphthyl ethylamine, r-+-a-1-naphthyl ethylamine, 1r-1-naphthalen-1-yl ethanamine
PubChem CID 2724264
IUPAC Name (1R)-1-naphthalen-1-ylethanamine
SMILES CC(C1=CC=CC2=CC=CC=C21)N

Specifications

Color Yellow
Boiling Point 80°C
Quantity 1 g
Formula Weight 171.24
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(+)-1-(1-Naphthyl)ethylamine
EINECSNumber (4)-1343
RTECSNumber QJ6963000
TSCA Yes
Recommended Storage Refrigerator