Learn More
(R)-(+)-1-(1-Naphthyl)ethylamine, 99%
CAS: 3886-70-2 | C12H13N | 171.243 g/mol
Supplier: Thermo Scientific Chemicals A1691814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (R)-(+)-1-(1-Naphthyl)ethylamine | |
| 1.06 | |
| >110°C (230°F) | |
| C12H13N | |
| MFCD00064114 | |
| UN2922 | |
| Air sensitive | |
| RTCUCQWIICFPOD-SECBINFHSA-N | |
| (1R)-1-naphthalen-1-ylethanamine | |
| 2724264 | |
| 99% |
| 3886-70-2 | |
| 152°C to 153°C (11 mmHg) | |
| Amine-like | |
| 1.623 | |
| 25 g | |
| 2208025 | |
| r-+-1-1-naphthyl ethylamine, r-1-naphthalen-1-yl ethanamine, 1r-1-naphthalen-1-yl ethan-1-amine, r-1-1-naphthyl ethylamine, 1r-1-naphthalen-1-ylethanamine, 1r-1-naphthylethylamine, r-+-alpha-1-naphthyl ethylamine, 1r-1-1-naphthyl ethylamine, r-+-a-1-naphthyl ethylamine, 1r-1-naphthalen-1-yl ethanamine | |
| CC(C1=CC=CC2=CC=CC=C21)N | |
| 171.243 | |
| 171.24 |
Chemical Identifiers
| 3886-70-2 | |
| 171.243 | |
| RTCUCQWIICFPOD-SECBINFHSA-N | |
| 2724264 | |
| CC(C1=CC=CC2=CC=CC=C21)N |
| C12H13N | |
| MFCD00064114 | |
| r-+-1-1-naphthyl ethylamine, r-1-naphthalen-1-yl ethanamine, 1r-1-naphthalen-1-yl ethan-1-amine, r-1-1-naphthyl ethylamine, 1r-1-naphthalen-1-ylethanamine, 1r-1-naphthylethylamine, r-+-alpha-1-naphthyl ethylamine, 1r-1-1-naphthyl ethylamine, r-+-a-1-naphthyl ethylamine, 1r-1-naphthalen-1-yl ethanamine | |
| (1R)-1-naphthalen-1-ylethanamine |
Safety and Handling
GHS H Statement
H301-H314-H318-H312
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 223-425-5
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only