missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-(1-Naphthyl)ethyl Isocyanate 90.0+%, TCI America™

Catalog No. I03361G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03361G 1 g
I03365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03361G Supplier TCI America Supplier No. I03361G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 42340-98-7
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00004015
InChI Key GONOHGQPZFXJOJ-SNVBAGLBSA-N
Synonym r---1-1-naphthyl ethyl isocyanate, isocyanic acid r---1-1-naphthyl ethyl ester, 1-1r-1-isocyanatoethyl naphthalene, naphthalene,1-1r-1-isocyanatoethyl, 1r-1-naphthylethanisocyanate, 1-1-isocyanatoethyl naphthalene #, pubchem8100, r-nei, r-1-1-naphthyl ethyl isocyanate
PubChem CID 170633
IUPAC Name 1-[(1R)-1-isocyanatoethyl]naphthalene
SMILES C[C@@H](N=C=O)C1=C2C=CC=CC2=CC=C1

Specifications

Color Yellow
Quantity 1 g
UN Number 2206
Formula Weight 197.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(-)-1-(1-Naphthyl)ethyl Isocyanate
TSCA No
Recommended Storage Refrigerator