missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl Hydrogen Phosphate 98.0+%, TCI America™
Supplier: TCI America B11435G
Specifications
| (R)-(-)-1,1′-Binaphthyl-2,2′-diyl Hydrogen Phosphate | |
| White-Yellow | |
| MFCD00010045 | |
| 1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, s-+-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, r---1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r---bnp acid, 11bs-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, r---1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, s-+-bnp acid, 4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide | |
| [O-]P1(=O)OC2=CC=C3C=CC=CC3=C2C2=C3C=CC=CC3=CC=C2O1 | |
| 347.29 | |
| 348.29 | |
| Crystalline Powder |
| 39648-67-4 | |
| C20H12O4P | |
| 5 g | |
| JEHUZVBIUCAMRZ-UHFFFAOYSA-M | |
| 13-oxo-12,14-dioxa-13λâµ-phosphapentacyclo[13.8.0.0²,¹¹.0³,â¸.0¹â¸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-olate | |
| 99589 | |
| ≥98.0% (T) |
Chemical Identifiers
| 39648-67-4 | |
| 347.29 | |
| JEHUZVBIUCAMRZ-UHFFFAOYSA-M | |
| 99589 | |
| [O-]P1(=O)OC2=CC=C3C=CC=CC3=C2C2=C3C=CC=CC3=CC=C2O1 |
| C20H12O4P | |
| MFCD00010045 | |
| 1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, s-+-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, r---1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r---bnp acid, 11bs-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, r---1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, s-+-bnp acid, 4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide | |
| 13-oxo-12,14-dioxa-13λâµ-phosphapentacyclo[13.8.0.0²,¹¹.0³,â¸.0¹â¸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-olate |
Safety and Handling
TSCA : No