Learn More
Selleck Chemical Llc QUIZARTINIB (AC220) 10MM/1ML DFS Item
Supplier:  Selleck Chemicals Co Ltd S152610MM1ML
Quizartinib (AC220) is a second-generation FLT3 inhibitor for Flt3(ITD/WT) with IC50 of 1.1 nM/4.2 nM in MV4-11 and RS4;11 cells, respectively, 10-fold more selective for Flt3 than KIT, PDGFRα, PDGFRβ, RET, and CSF-1R. Phase 3.
Catalog No. NC1323520
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions