missing translation for 'onlineSavingsMsg'
Learn More

Quinuclidine 96.0+%, TCI America™

Catalog No. p-7135634
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q0062200MG 200 mg
Q00621G 1 g
Q00625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. Q0062200MG Supplier TCI America Supplier No. Q0062200MG
Only null left

Chemical Identifiers

CAS 100-76-5
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.188
MDL Number MFCD00006690
InChI Key SBYHFKPVCBCYGV-UHFFFAOYSA-N
Synonym quinuclidine, 1-azabicyclo 2.2.2 octane, 1,4-ethanopiperidine, 1,4-ethylenepiperidine, chinuclidin, abco, unii-xfx99fc5vi, 4-azabicyclo 2.2.2 octane, xfx99fc5vi
PubChem CID 7527
ChEBI CHEBI:38420
IUPAC Name 1-azabicyclo[2.2.2]octane
SMILES C1CN2CCC1CC2

Specifications

Color White-Yellow
Quantity 200 mg
UN Number 2811
Formula Weight 111.19
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Quinuclidine
RTECSNumber CL5594625
TSCA Yes