missing translation for 'onlineSavingsMsg'
Learn More

Quinuclidine Hydrochloride 98.0+%, TCI America™

Catalog No. Q00501G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00501G 1 g
Q00505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00501G Supplier TCI America Supplier No. Q00501G

Chemical Identifiers

CAS 39896-06-5
Molecular Formula C7H14ClN
Molecular Weight (g/mol) 147.646
MDL Number MFCD00012727
InChI Key BZLBBZLOMXKMTA-UHFFFAOYSA-N
Synonym 1-Azabicyclo[2.2.2]octane Hydrochloride
PubChem CID 520969
IUPAC Name 1-azabicyclo[2.2.2]octane;hydrochloride
SMILES C1CN2CCC1CC2.Cl

Specifications

Melting Point 365°C
Color White
Quantity 1 g
Formula Weight 147.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Quinuclidine Hydrochloride