missing translation for 'onlineSavingsMsg'
Learn More

Quinoxaline, 98+%, Thermo Scientific Chemicals

Catalog No. p-3758740
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC132221000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC132221000 Supplier Thermo Scientific Chemicals Supplier No. 132221000
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91-19-0
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00006719
InChI Key XSCHRSMBECNVNS-UHFFFAOYSA-N
Synonym benzoparadiazine, 1,4-benzodiazine, phenopiazine, phenpiazine, quinazine, benzopyrazine, 1,4-diazanaphthalene, benzo a pyrazine, 1,4-naphthyridine, chinoxalin
PubChem CID 7045
ChEBI CHEBI:36616
IUPAC Name quinoxaline
SMILES C1=CC=C2C(=C1)N=CC=N2

Specifications

Melting Point 29°C to 34°C
Color Yellow to Brown
Boiling Point 220°C to 223°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 23, 176
Merck Index 15, 8195
Solubility Information Solubility in water: soluble. Other solubilities: miscible in alcohol,ether,benzene,acetone
Formula Weight 130.15
Percent Purity 98+%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Quinoxaline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 202-047-4

RUO – Research Use Only