missing translation for 'onlineSavingsMsg'
Learn More

Quinoxaline, 98+%

CAS: 91-19-0 | C8H6N2 | 130.15 g/mol

Supplier:  Thermo Scientific Chemicals A1405522

 View more versions of this product

Catalog No. AAA1405522


Only null left
Explore available promotions

Description

Description

Used as dyes, pharmaceuticals and antibiotics intermediate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Quinoxaline
27°C to 32°C
220°C to 223°C
C8H6N2
100 g
14,8078
XSCHRSMBECNVNS-UHFFFAOYSA-N
quinoxaline
7045
130.15
91-19-0
1.124
98°C (208°F)
MFCD00006719
109351
benzoparadiazine, 1,4-benzodiazine, phenopiazine, phenpiazine, quinazine, benzopyrazine, 1,4-diazanaphthalene, benzo a pyrazine, 1,4-naphthyridine, chinoxalin
C1=CC=C2C(=C1)N=CC=N2
130.15
CHEBI:36616
≥98%

Chemical Identifiers

91-19-0
130.15
XSCHRSMBECNVNS-UHFFFAOYSA-N
7045
quinoxaline
C8H6N2
MFCD00006719
benzoparadiazine, 1,4-benzodiazine, phenopiazine, phenpiazine, quinazine, benzopyrazine, 1,4-diazanaphthalene, benzo a pyrazine, 1,4-naphthyridine, chinoxalin
CHEBI:36616
C1=CC=C2C(=C1)N=CC=N2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 202-047-4

RTECSNumber : VD1225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only