missing translation for 'onlineSavingsMsg'
Learn More

Quinoxaline-6-carboxylic acid, 95%

Catalog No. AAH3315703
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3315703 1 g
AAH3315706 5 g
AAH3315714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3315703 Supplier Thermo Scientific Chemicals Supplier No. H3315703

CAS: 6925-00-4 | C9H6N2O2 | 174.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6925-00-4
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
MDL Number MFCD01365834
InChI Key JGQDBVXRYDEWGM-UHFFFAOYSA-N
Synonym 6-quinoxalinecarboxylic acid, chinoxalin-6-carbonsaeure, quinoxaline-6-carboxylicacid, pubchem15419, quinoxaline-6-carboxylic acid
PubChem CID 674813
IUPAC Name quinoxaline-6-carboxylic acid
SMILES C1=CC2=NC=CN=C2C=C1C(=O)O

Specifications

Melting Point 248°C to 251°C
Quantity 1 g
Formula Weight 174.16
Percent Purity 95%
Chemical Name or Material Quinoxaline-6-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only